missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klercide™ 70/30 Denatured Ethanol

For use in effective disinfection procedure. Ecolab™ Klercide™ 70/30 Denatured Ethanol is sterilised by gamma irradiation.

Brand:  Ecolab™ 3078260

704.81 GBP valid until 2025-05-02
Use promo code "24151" to get your promotional price.



 View more versions of this product

Product Code. 15356068

  • £949.18 / Pack of 12

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Available as either 70/30 Isopropyl Alcohol (IPA) or Denatured Ethanol blended with water for injection (WFI) or deionized water
  • Also available as 60% Isopropyl alcohol or Propan-1-ol and Denatured Ethanol, both blended with WFI
  • Use pharma grade alcohol in blend
  • Cleanroom processed, 0.2 micron filtered and terminally sterilized by gamma irradiation
  • Available as spray systems fitted with SteriShield Delivery System, which ensures sterility of contents is preserved throughout use, aerosols, stream aerosol or in 5L capped container
  • Available with complete supporting documentation
Specifications

Specifications

Alcohol
Filtered
500 mL
Sterile
Effective disinfection procedure
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Klercide 70/30 Denatured Ethanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, 2-methylpropan-2-ol

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ecolab™ Klercide™ 70/30 Denatured Ethanol > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.