Learn More
Thermo Scientific™ Elite pH Pocket Testers and Replacement Sensors
Flexible on-the-go pH and temperature testing using durable Elite pocket testers with replaceable sensors for a variety of applications including environmental monitoring, pools, spas and more!
£39.92 - £149.17
Product Code | Brand | Description | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Price | Quantity & Availability | |||||
16846966
|
Thermo Scientific™
01X001535 |
Replacement sensor for Elite pH pocket tester | 1 replacement pH sensor |
£39.92
Each |
In Stock
Log in to see stock. |
|||||
15914022
|
Thermo Scientific™
ELITEPHKIT |
Elite pH pocket tester for measuring pH with temperature | Elite pH tester with batteries, lanyard, soft carry case and pH buffers |
£149.17
Each |
In Stock
Log in to see stock. ![]() |
|||||
15904022
|
Thermo Scientific™
ELITEPH |
Elite pH pocket tester for measuring pH with temperature | Elite pH tester with batteries, lanyard and soft carry case |
£125.44
Each |
In Stock
Log in to see stock. |
|||||
Description
Benefit from these pocket-sized testers. Thermo Scientific™ Elite testers are designed to provide on-the-go measurements for a variety of applications including homebrew beer and wine, food and beverage, hydroponics, aquaculture and aquariums, agriculture, pools and spas, environmental monitoring, education and more. The Elite CTS cup-sensor pocket testers are ideal for flexible on-the-go conductivity, TDS, salinity and temperature testing in a compact, travel-ready design.
Elite Pocket Testers offer flexible on-the-go electrochemical testing. Choose the Elite pH model to measure pH with temperature using a direct-measure sensor design for your application. Elite pocket testers are durable, with a large backlit screen, one-touch calibration feature and replaceable sensor heads. They are ideal for testing beverages including homebrew beer and wine, food, hydroponics, aquaculture and aquariums, environmental monitoring and more.
- Flexible testing option
- Measure pH and temperature
- Large back-lit graphic display
- Uses 4 x AAA alkaline batteries
- Average 250 hours of battery life
- Wide leak-proof cap serves as stable base
- Replaceable sensors
- One-year warranty for tester
- Six-month warranty for replacement sensors