Learn More
Thermo Scientific™ Elite pH Pocket Testers and Replacement Sensors
Flexible on-the-go pH and temperature testing using durable Elite pocket testers with replaceable sensors for a variety of applications including environmental monitoring, pools, spas and more!
Brand: Thermo Scientific™ ELITEPHKIT
86.94 GBP valid until 2025-04-18
Use promo code "24095" to get your promotional price.
Description
Benefit from these pocket-sized testers. Thermo Scientific™ Elite testers are designed to provide on-the-go measurements for a variety of applications including homebrew beer and wine, food and beverage, hydroponics, aquaculture and aquariums, agriculture, pools and spas, environmental monitoring, education and more. The Elite CTS cup-sensor pocket testers are ideal for flexible on-the-go conductivity, TDS, salinity and temperature testing in a compact, travel-ready design.
Elite Pocket Testers offer flexible on-the-go electrochemical testing. Choose the Elite pH model to measure pH with temperature using a direct-measure sensor design for your application. Elite pocket testers are durable, with a large backlit screen, one-touch calibration feature and replaceable sensor heads. They are ideal for testing beverages including homebrew beer and wine, food, hydroponics, aquaculture and aquariums, environmental monitoring and more.
- Flexible testing option
- Measure pH and temperature
- Large back-lit graphic display
- Uses 4 x AAA alkaline batteries
- Average 250 hours of battery life
- Wide leak-proof cap serves as stable base
- Replaceable sensors
- One-year warranty for tester
- Six-month warranty for replacement sensors

Specifications
Elite pH pocket tester for measuring pH with temperature | |
±0.6°C/±1.0°F for 0.0deg;C to 50.0°C | |
pH, Temperature | |
-1 to 15 | |
1 to 3 | |
32.0°F to 140.0°F | |
1 point | |
Yes | |
IP-67 | |
5 to 85 %, non-condensing | |
Yes | |
4 x AAA Alkaline |
Elite pH tester with batteries, lanyard, soft carry case and pH buffers | |
±0.02 | |
pH, temperature | |
0.01 | |
0°C to 60°C | |
0.1°C, 0.1°F | |
Graphic Dot Matrix | |
CE, FCC | |
1 Year for Tester | |
Stability icon on display | |
5°C to 45°C (ambient) | |
250 hr. (average) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.