Learn More
Elma™ Elma Dry
The Elma Dry allows cleaned items to drip dry or cool down and dry as the final step in the cleaning process.
Brand:  Elma™ 1078542
 View more versions of this product
Product Code. 17831727
  • £474.00 / Each
Estimated Shipment: 14-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Elma Dry units are available in three sizes (30, 120 and 300). They are designed for use in production facilities, workshops and service areas.
  • After cleaning in the ultrasonic bath, the items in the Elma™ basket are first rinsed in the Elma™ Sink and then dried easily and conveniently in the Elma Dry
  • Cold/hot-air blower (70°C) ensures that even delicate parts can be dried gently
  • In cold-air mode, the parts can also be cooled down quickly
  • Cover includes ventilation slits that maximize the air turbulence and discharge the humid air to the outside while reducing energy consumption
  • Dust filter on the underside of the base is replaceable and prevents recontamination of the cleaned parts due to dust in the room air
  • Due to the safety shutdown when the limit temperature is reached, Elma Dry units are both safe and gentle on their contents
Specifications
Specifications
Elma Dry
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
TD 120