Learn More
Elma™ Elma Dry
The Elma Dry allows cleaned items to drip dry or cool down and dry as the final step in the cleaning process.
£336.00 - £855.00
Specifications
Type | Elma Dry |
---|---|
For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
17851727
|
Elma™
1082175 |
TD 30 |
£336.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
17831727
|
Elma™
1078542 |
TD 120 |
£474.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
17821727
|
Elma™
1078575 |
TD 300 |
£855.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
Descripción
Elma Dry units are available in three sizes (30, 120 and 300). They are designed for use in production facilities, workshops and service areas.
- After cleaning in the ultrasonic bath, the items in the Elma™ basket are first rinsed in the Elma™ Sink and then dried easily and conveniently in the Elma Dry
- Cold/hot-air blower (70°C) ensures that even delicate parts can be dried gently
- In cold-air mode, the parts can also be cooled down quickly
- Cover includes ventilation slits that maximize the air turbulence and discharge the humid air to the outside while reducing energy consumption
- Dust filter on the underside of the base is replaceable and prevents recontamination of the cleaned parts due to dust in the room air
- Due to the safety shutdown when the limit temperature is reached, Elma Dry units are both safe and gentle on their contents
Especificaciones
Elma Dry | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |