missing translation for 'onlineSavingsMsg'
Learn More

Merck Centricon™ Plus-70 Centrifugal Filters

15 mL to 70 mL volume

Brand:  Merck UFC710008

251.56 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 72

Additional Details : Weight : 0.61380kg

 View more versions of this product

Product Code. 10654923

  • £429.00 / Pack of 8
Estimated Shipment: 19-08-2024
to see stock.
Add to basket

Description

Description

The Centricon™ Plus-70 centrifugal filter is a disposable, single-use device designed for rapid processing of aqueous biological solutions in volumes ranging from 15 to 70 mL. It is compatible with swinging-bucket centrifuges only. The Centricon Plus-70 device can concentrate most 70 mL solutions down to 350 μL in just 25 minutes (some solutions may take slightly longer).•90% typical recovery •50X to 200X concentration •Low hold-up volume •Polypropylene housing minimizes binding •Deadstop prevents spinning to dryness •Ready to use. Nothing to assemble Applications:•Concentrating and desalting chromatography column eluates •Concentrating monoclonal antibodies •Concentrating proteins or viruses from culture supernatants •Clarifying tissue homogenates and cell lysates •Buffer exchange or diafiltration

disposable,single-use,aqueous,solutions,volumes,ranging

Specifications

Specifications

Single-use
100 kDa
Concentrating and desalting chromatography column eluates; Concentrating monoclonal antibodies; Concentrating proteins or viruses from culture supernatants; Clarifying tissue homogenates and cell lysates; Buffer exchange or diafiltration
Polypropylene filtrate collection cup; sample filter cup; concentrate/retentate cup and cap
15 to 70 mL
Ultracel-PL
Non-sterile
Regenerated Cellulose (RC)
Centrifugal Filters
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers