missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Centricon™ Plus-70 Centrifugal Filters
15 mL to 70 mL volume
£313.00 - £429.00
Specifications
Disposable | Single-use |
---|---|
Sterility | Non-sterile |
Housing Material | Regenerated Cellulose (RC) |
Type | Centrifugal Filters |
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
10296062
|
Merck
UFC701008 |
10 kDa |
£319.00
Pack of 8 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
10654923
|
Merck
UFC710008 |
100 kDa |
£429.00
Pack of 8 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
10042002
|
Merck
UFC703008 |
30 kDa |
£313.00
Pack of 8 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
12971721
|
Merck
UFC700308 |
3 kDa |
£340.00
Pack of 8 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
Description
The Centricon™ Plus-70 centrifugal filter is a disposable, single-use device designed for rapid processing of aqueous biological solutions in volumes ranging from 15 to 70 mL. It is compatible with swinging-bucket centrifuges only. The Centricon Plus-70 device can concentrate most 70 mL solutions down to 350 μL in just 25 minutes (some solutions may take slightly longer).•90% typical recovery •50X to 200X concentration •Low hold-up volume •Polypropylene housing minimizes binding •Deadstop prevents spinning to dryness •Ready to use. Nothing to assemble Applications:•Concentrating and desalting chromatography column eluates •Concentrating monoclonal antibodies •Concentrating proteins or viruses from culture supernatants •Clarifying tissue homogenates and cell lysates •Buffer exchange or diafiltrationdisposable,single-use,aqueous,solutions,volumes,ranging
Specifications
Single-use | |
Non-sterile | |
Regenerated Cellulose (RC) | |
Centrifugal Filters |