missing translation for 'onlineSavingsMsg'
Learn More

Merck Centricon™ Plus-70 Centrifugal Filters

15 mL to 70 mL volume

Brand:  Merck UFC700308

274.53 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 72

Additional Details : Weight : 0.58500kg

 View more versions of this product

Product Code. 12971721

  • £340.00 / Pack of 8
Estimated Shipment: 29-07-2024
to see stock.
Add to basket

Description

Description

The Centricon™ Plus-70 centrifugal filter is a disposable, single-use device designed for rapid processing of aqueous biological solutions in volumes ranging from 15 to 70 mL. It is compatible with swinging-bucket centrifuges only. The Centricon Plus-70 device can concentrate most 70 mL solutions down to 350 μL in just 25 minutes (some solutions may take slightly longer).•90% typical recovery •50X to 200X concentration •Low hold-up volume •Polypropylene housing minimizes binding •Deadstop prevents spinning to dryness •Ready to use. Nothing to assemble Applications:•Concentrating and desalting chromatography column eluates •Concentrating monoclonal antibodies •Concentrating proteins or viruses from culture supernatants •Clarifying tissue homogenates and cell lysates •Buffer exchange or diafiltration

disposable,single-use,aqueous,solutions,volumes,ranging

Specifications

Specifications

Single-use
3 kDa
19 cm2
Regenerated Cellulose (RC)
15 to 70 mL
Polypropylene
Non-sterile
For concentration, purification of biological sample; for rapid processing of aq. Soln.
Centrifugal Filters
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers