missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Immobilon™ Western Chemiluminescent HRP Substrate (ECL)
Provides high sensitivity over a broad detection range at an economical price
Brand: Merck WBKLS0050
79.96 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Includes: Luminol, peroxide solution
Description
- Combines high sensitivity and low background
- Allows detection of high- and low-abundance proteins in the mid-femtogram range
- Lowers antibody consumption by two- to five-fold compared to less sensitive reagents
- Compatible with PVDF and nitrocellulose membranes as well as all commonly used buffers and blocking reagents
Specifications
Dot/Slot Blot | |
Horseradish Peroxidase | |
25mL luminol, 25mL peroxide solution |
2 x 25 mL | |
500cm2 Membrane | |
2°C to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction