missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Immobilon™ Western Chemiluminescent HRP Substrate (ECL)
Provides high sensitivity over a broad detection range at an economical price
£145.00 - £621.00
Specifications
For Use With (Application) | Dot/Slot Blot |
---|---|
Substrate | Horseradish Peroxidase |
Content And Storage | 2°C to 8°C |
Includes: Luminol, peroxide solution
Product Code | Brand | Kit Contents | Quantity | Sufficient For | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Kit Contents | Quantity | Sufficient For | Price | Quantity & Availability | |||
11536345
|
Merck
WBKLS0050 |
25mL luminol, 25mL peroxide solution | 2 x 25 mL | 500cm2 Membrane |
£145.00
Pack of 2 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11556345
|
Merck
WBKLS0500 |
250mL luminol, 250mL peroxide solution | 2 x 250 mL | 5000cm2 Membrane |
£621.00
Pack of 2 |
In Stock
Log in to see stock. |
|||
11546345
|
Merck
WBKLS0100 |
50mL luminol, 50mL peroxide solution | 2 x 50 mL | 1000cm2 Membrane |
£243.00
Pack of 2 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
Description
- Combines high sensitivity and low background
- Allows detection of high- and low-abundance proteins in the mid-femtogram range
- Lowers antibody consumption by two- to five-fold compared to less sensitive reagents
- Compatible with PVDF and nitrocellulose membranes as well as all commonly used buffers and blocking reagents
Specifications
Dot/Slot Blot | |
Horseradish Peroxidase | |
2°C to 8°C |