missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon™ Western Chemiluminescent HRP Substrate (ECL)

Provides high sensitivity over a broad detection range at an economical price

Brand:  Merck WBKLS0100

139.29 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11546345

  • £298.00 / Pack of 2
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Luminol, peroxide solution
This item is not returnable. View return policy

Description

Description

  • Combines high sensitivity and low background
  • Allows detection of high- and low-abundance proteins in the mid-femtogram range
  • Lowers antibody consumption by two- to five-fold compared to less sensitive reagents
  • Compatible with PVDF and nitrocellulose membranes as well as all commonly used buffers and blocking reagents

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dot/Slot Blot
Horseradish Peroxidase
50mL luminol, 50mL peroxide solution
2 x 50 mL
1000cm2 Membrane
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.