Learn More
Merck Replacement Electrodes for Millicell-ERS Volt-Ohm Meter, 1 pair
Measures Trans Epithelial Electrical Resistance (TEER) of epithelial cells in culture
Brand:  Merck MERSSTX01
Product Code. 10517682
  • £547.00 / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
Description
Description
Provides qualitative measurement of cell monolayer health and quantative measures of confluency. Designed for use with cells cultured in Millicell™ Culture Plate Inserts only.
Utilizes AC current for membrane resistance measurements, producing a zero net charge on cells and eliminating adverse DC current effects on cell membranes. A silver/silver chloride (Ag/AgCI) pellet on each electrode tip measures voltage. Due to the small size of the electrodes, the user can easily measure transepithelial voltage and the resistance of cells grown on microporous membranes.
Services
Electrodes and batteries are warranted for 30 days only from the date on which the customer receives these items
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Replacement Electrode
Millicell-ERS Volt-Ohm Meter
To provides qualitative measurement of cell monolayer health and quantative measures of confluency
12.5 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Merck Replacement Electrodes for Millicell-ERS Volt-Ohm Meter, 1 pair > Replacement Electrodes