missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipak™ Disposable Filter Units

Removes particles and microorganisms from liquids and gases with pump-driven filter. Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.

Brand:  Merck Chemicals MPVL02GH2

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 17240794

  • £282.00 / Each
Estimated Shipment: 08-10-2024
to see stock.
Add to basket

Description

Description

Removes particles and microorganisms from liquids and gases with pump-driven filter. Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.

Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.

  • Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
  • Point-of-use filter for high-purity water
  • Ideally suited for high value-added applications
  • Durapore™ PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
  • Presterilized units are gamma irradiated
  • Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
  • 100% integrity tested to guarantee integrity

Specifications

Specifications

0.25 in./6.35mm stepped hose barb inlet and outlet with bell
Polycarbonate
0.1 μm
100 cm2
Removal of particles and microorganisms from liquids and gases
2/Pk.
10 L
8.4 cm
7.6 cm
Single-use
PVDF
Sterile
0.7 L/min. at 1.75 bar
75 psi at 25°C
Filter Unit
3.3 in.
3 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers