missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millipak™ Disposable Filter Units
Removes particles and microorganisms from liquids and gases with pump-driven filter. Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.
£259.00 - £507.00
Specifications
Disposable | Single-use |
---|---|
Material | Polycarbonate |
Max. Inlet Pressure | 75 psi at 25°C |
Type | Filter Unit |
Diameter (English) | 3 in. |
Product Code | Brand | Connectors | Flow Rate | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connectors | Flow Rate | Volume (Metric) | Price | Quantity & Availability | |||
12447146
|
Merck
MPGL06GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 3.8 L/min. at 1.75 bar | 30 L |
£399.00
Pack of 2 |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||
17848973
|
Merck
MPHL04GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 8.5 L/min. at 2.45 bar | 20 L |
£311.00
Pack of 2 |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16632782
|
Merck
MPHL02GH2 |
0.25 in. /6.35mm stepped hose barb inlet and outlet with bell | 6.4 L/min. at 2.45 bar | 10 L |
£259.00
Pack of 2 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17240794
|
Merck Chemicals
MPVL02GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 0.7 L/min. at 1.75 bar | 10 L |
£282.00
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||
12457146
|
Merck
MPVL06GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 1.9 L/min. at 1.75 bar | 30 L |
£507.00
Pack of 2 |
N/A | |||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
17250794
|
Merck
MPVL06GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 1.9 L/min. at 1.75 bar | 30 L |
£439.00
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
||
Description
Removes particles and microorganisms from liquids and gases with pump-driven filter. Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.
Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.
- Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
- Point-of-use filter for high-purity water
- Ideally suited for high value-added applications
- Durapore™ PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
- Presterilized units are gamma irradiated
- Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
- 100% integrity tested to guarantee integrity
Specifications
Single-use | |
75 psi at 25°C | |
3 in. |
Polycarbonate | |
Filter Unit | |
7.6 cm |