Learn More
Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
Brand:  Merck MSIPN4W50
 View more versions of this product
Product Code. 10307473
  • £1865.00 / Pack of 50
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
MultiScreen-IP Filter Plate with Immobilon™-P membrane
0.45 μm
ABS Plastic
Non-sterile
ELISPOT assays
Hydrophobic PVDF
8 x 12
White
96
0.28cm2
50/Pk.