Learn More
Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
Brand:  Merck MSIPS4W10
Code : 73
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10402362
  • £464.00 / Pack of 10
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
Specifications
Specifications
Standard
MultiScreen™HTS-IP plate with Immobilon™-P membrane
0.45 μm
MultiScreenHTS-IP Sterile Filter Plate
96
0.28cm2
10/Pk.
Hydrophobic PVDF
8 x 12
White
Acrylic
Sterile
ELISPOT assays
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificats
Voir les accessoires