Learn More
Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
Brand:  Merck MAHAS4510
 View more versions of this product
Product Code. 10258100
  • £482.00 / Pack of 10
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
MultiScreen™-HA with MCE membrane
0.45 μm
Styrene
Sterile
ELISPOT assays
Mixed Cellulose Esters
8 x 12
Clear
96
0.28 cm2
10/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products