missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ 96-Well Assay Systems
Perform cell growth, feeding, and analysis in one plate
Brand: Merck PSRP004R1
This item is not returnable.
View return policy
Description
- Patented 96-well device supports epithelial cell growth and differentiation of the Caco-2 and other cell lines
- After the formation of a differentiated Caco-2 monolayer, the assembly can be used to measure the rate of drug transport across the Caco-2 cell barrier
- All procedures are designed to be carried out in a single device
- Apical and basolateral access ports allow contamination-free access to cells, easy feeding and media changes, and effective sample analysis
- Increased sample throughput when compared to 24-well systems
- Use manually or with automated systems
- Versatile format with 96- or single-well feeder trays
Specifications
Standard | |
Polyethylene Terephthalate | |
Individually Wrapped | |
Clear | |
Polystyrene | |
Sterile | |
Cell attachment, growth and differentiation for many cell applications | |
Each | |
1.28 cm | |
2.62 cm |
With Lid | |
8 x 12 | |
1 μm | |
Millicell 96 Cell Culture Plate | |
96 | |
0.12 cm2 | |
96-well cell culture plate, single well feeder tray, 96-well receiver tray, lid | |
0.5 x 3.33 x 1.03 in. (1.28 x 8.48 x 2.62 cm) | |
8.48 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction