missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ 96-Well Assay Systems
Perform cell growth, feeding, and analysis in one plate
£680.00 - £3110.00
Specifications
Format | Standard |
---|---|
Lid | With Lid |
Membrane | Polyethylene Terephthalate |
Array | 8 x 12 |
Pore Size | 1 μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10727094
|
Merck
PSRP004R5 |
5/Pk. |
£3110.00
Pack of 5 |
Estimated Shipment: 24-12-2024
Log in to see stock. |
|||||
10489173
|
Merck
PSRP004R1 |
Each |
£680.00
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
Description
- Patented 96-well device supports epithelial cell growth and differentiation of the Caco-2 and other cell lines
- After the formation of a differentiated Caco-2 monolayer, the assembly can be used to measure the rate of drug transport across the Caco-2 cell barrier
- All procedures are designed to be carried out in a single device
- Apical and basolateral access ports allow contamination-free access to cells, easy feeding and media changes, and effective sample analysis
- Increased sample throughput when compared to 24-well systems
- Use manually or with automated systems
- Versatile format with 96- or single-well feeder trays
Specifications
Standard | |
Polyethylene Terephthalate | |
1 μm | |
Polystyrene | |
Sterile | |
Cell attachment, growth and differentiation for many cell applications | |
0.5 x 3.33 x 1.03 in. (1.28 x 8.48 x 2.62 cm) | |
8.48 cm |
With Lid | |
8 x 12 | |
Clear | |
96 | |
0.12 cm2 | |
96-well cell culture plate, single well feeder tray, 96-well receiver tray, lid | |
1.28 cm | |
2.62 cm |