Learn More
Merck MultiScreen™ HTS 96-Well Filter Plates with MCE Membrane
Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies
Brand:  Merck MSHAS4510
Code : 73
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10317183
  • £398.00 / Pack of 10
Estimated Shipment: 06-08-2024
to see stock.
Add to basket
Description
Description
  • Developed for high-throughput use with automated work stations
  • Individually sealed wells prevent incubation crosstalk
  • Removable underdrain allows access to the filters
  • Rigid sidewalls improve handling
  • Ample surfaces provided for bar code labels
  • Pore size: .45μm
Solid-phase RIA, DNA hybridization, ELISA and protein binding
Specifications
Specifications
Standard
8 x 12
Clear
96
0.28 cm2
10/Pk.
Mixed Cellulose Esters
0.45 μm
Styrene
Sterile
Solid phase RIA, DNA hybridization, ELISA, protein binding
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products