missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ HTS 96-Well Filter Plates with MCE Membrane
Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies
£398.00 - £1940.00
Specifications
Format | Standard |
---|---|
Membrane | Mixed Cellulose Esters |
Array | 8 x 12 |
Pore Size | 0.45 μm |
No. of Wells | 96 |
Product Code | Brand | Color | Material | Sterility | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | Sterility | Quantity | Price | Quantity & Availability | ||
10184073
|
Merck
MSHAN4510 |
Clear | Styrene | Non-sterile | 10/Pk. |
£421.00
Pack of 10 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||
10689113
|
Merck
MSHAN4550 |
Clear | Styrene | Non-sterile | 50/Pk. |
£1700.00
Pack of 50 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
||
10307183
|
Merck
MSHAN4B50 |
Opaque | Barex | Non-sterile | 50/Pk. |
£1940.00
Pack of 50 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
||
10317183
|
Merck
MSHAS4510 |
Clear | Styrene | Sterile | 10/Pk. |
£398.00
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
10582264
|
Merck
MSHAS4B10 |
Opaque | Barex | Sterile | 10/Pk. |
£496.00
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
Description
- Developed for high-throughput use with automated work stations
- Individually sealed wells prevent incubation crosstalk
- Removable underdrain allows access to the filters
- Rigid sidewalls improve handling
- Ample surfaces provided for bar code labels
- Pore size: .45μm
Solid-phase RIA, DNA hybridization, ELISA and protein binding
Specifications
Standard | |
8 x 12 | |
96 |
Mixed Cellulose Esters | |
0.45 μm | |
Solid phase RIA, DNA hybridization, ELISA, protein binding |