Learn More
Merck MultiScreen™ Sealing Tape
Choose from opaque or clear
Brand:  Merck MATAH0P00
Code : 72
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11758522
  • £117.00 / Pack of 100
Estimated Shipment: 13-08-2024
to see stock.
Add to basket
Description
Description
  • Opaque sealing tape is used to seal 100μL plate bottoms read in TopCount™ instruments
  • Clear sealing tape can be used to seal all manufacturers' 300μL plate tops and plate bottoms for MicroBeta™ readers
  • Precut to size
Specifications
Specifications
Opaque
100
Sealing Tape
Non-sterile
Used to seal 100μL plate bottoms read in TopCount™ instruments