Learn More
Merck MultiScreenHTS Solubility Filter Plate
96-well filter plate designed and optimized for aqueous solubility assays
Brand:  Merck MSSLBPC10
Code : 73
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10182792
  • £710.00 / Pack of 10
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • High-throughput system to classify or quantify the aqueous solubility of compound libraries stored in solvent as a concentrated solution
  • Analysis time in under four hours using <100μg of sample per test
  • Automation compatible, with space for bar coding
  • Validated for high drug recovery
  • Low-binding membrane and low-binding plate materials
Specifications
Specifications
Standard
8 x 12
Clear
96
Aqueous solubility assays
Polycarbonate
0.4 μm
Styrene
Non-sterile
10/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products