missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreenHTS Solubility Filter Plate
96-well filter plate designed and optimized for aqueous solubility assays
£710.00 - £3395.00
Specifications
Format | Standard |
---|---|
Membrane | Polycarbonate |
Array | 8 x 12 |
Pore Size | 0.4 μm |
Color | Clear |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10637303
|
Merck
MSSLBPC50 |
50/Pk. |
£3395.00
Pack of 50 |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10182792
|
Merck
MSSLBPC10 |
10/Pk. |
£710.00
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
- High-throughput system to classify or quantify the aqueous solubility of compound libraries stored in solvent as a concentrated solution
- Analysis time in under four hours using <100μg of sample per test
- Automation compatible, with space for bar coding
- Validated for high drug recovery Low-binding membrane and low-binding plate materials
Specifications
Standard | |
8 x 12 | |
Clear | |
96 | |
Aqueous solubility assays |
Polycarbonate | |
0.4 μm | |
Styrene | |
Non-sterile |