missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck PureProteome™ Nickel Magnetic Beads
Offer yields comparable with other magnetic purification systems while offering high purities
£185.00 - £825.00
Specifications
Binding Property | 1 to 5.5mg/mL |
---|---|
Content And Storage | 2°C to 8°C |
Particle Size | 10 μm |
For Use With (Application) | Purification of His-tagged recombinant Protein |
Type | Magnetic Affinity Bead |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11730353
|
Merck
LSKMAGH02 |
2 x 1 mL |
£185.00
Pack of 2 |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
11750343
|
Merck
LSKMAGH10 |
10 mL |
£825.00
Each |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||||
Description
- No lysate clearing—crude cell lysates are processed by magnetic beads
- Fast processing time—entire protocol is completed in less than one hour
- High capacity—bind 5 to 15mg of protein per mL of settled beads
- High affinity—binding can take place in buffer containing <5mM EDTA
- Flexible scale—varying the bead quantity allows different protein yields
- Flexible elution—native or denatured proteins
- Automatable—hands-free screening process
Specifications
1 to 5.5mg/mL | |
10 μm | |
Magnetic Affinity Bead |
2°C to 8°C | |
Purification of His-tagged recombinant Protein |