missing translation for 'onlineSavingsMsg'
Learn More

Merck Steripak™-GP Sterile Filter Units

Fast flow and high throughput

Brand:  Merck SPGPM10RJ

Code : 71

Additional Details : Weight : 0.47700kg

 View more versions of this product

Product Code. 10594512

  • £195.00 / Pack of 3
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Fast flow and high throughput

Steripak-GP filter units are designed for larger scale pressure-driven filtration of tissue culture media with or without serum. They are single use and are not autoclavable. The filter units come in two sizes, with the smaller unit able to filter up to 10 L and the larger up to 20 L. Both sizes utilize the low protein binding, fast flow 0.22 μm Millipore Express membrane. Each unit has an integral filling bell for bottle-top use and a manually operated upstream air vent to prevent membrane air locking. They are supplied sterile and ready to connect to a pump or pressure vessel (not included).

Steripak™-GP filter units are designed for larger scale, pressure-driven filtration of tissue culture media with or without serum. Each filter is for a single use and is not autoclavable. The filter units come in two sizes with the smaller unit able to filter up to 10 L and the larger up to 20 L.

Features & Benefits:
  • Millipore Express™ PLUS membrane for fastest flow rates and low protein binding
  • Larger scale, pressure-driven filtration device for faster flow rates speeds time-consuming, large-volume filtrations
  • 10 L and 20 L capacity sizes for maximum laboratory-scale filtration options
  • Manually operated top air vent to prevent air lock stop of flow
  • Filling bell for cleaner filling environment around the opening of the receptacle bottle

Applications:
Sterilization of Aqueous Solutions, Media Preparation, Buffer Preparation

Specifications

Specifications

Single-use
PES
Sterile
3Pack
6mm stepped hose barb, filling bell
45°C
10 mL
7.6 cm
SAN
0.22 μm
75 psi forward, 5 psi reverse
6 mm stepped hose barb, filling bell
113°F
Bottle Top Filter
2.99 in.
9.9 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers