missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Steripak™-GP Sterile Filter Units
Fast flow and high throughput
£195.00 - £247.00
Specifications
Disposable | Single-use |
---|---|
Material | SAN |
Material (Membrane) | PES |
Pore Size | 0.22 μm |
Sterility | Sterile |
Includes: 6mm (0.25 in. ) stepped hose barb inlet, 6mm (0.25 in. ) stepped hose barb outlet and filling bell.
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10594512
|
Merck
SPGPM10RJ |
10 mL |
£195.00
Pack of 3 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
10503994
|
Merck
SPGPM20RJ |
1 mL |
£247.00
Pack of 3 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
Description
Fast flow and high throughput
Steripak-GP filter units are designed for larger scale pressure-driven filtration of tissue culture media with or without serum. They are single use and are not autoclavable. The filter units come in two sizes, with the smaller unit able to filter up to 10 L and the larger up to 20 L. Both sizes utilize the low protein binding, fast flow 0.22 μm Millipore Express membrane. Each unit has an integral filling bell for bottle-top use and a manually operated upstream air vent to prevent membrane air locking. They are supplied sterile and ready to connect to a pump or pressure vessel (not included).Steripak™-GP filter units are designed for larger scale, pressure-driven filtration of tissue culture media with or without serum. Each filter is for a single use and is not autoclavable. The filter units come in two sizes with the smaller unit able to filter up to 10 L and the larger up to 20 L.
Features & Benefits:
- Millipore Express™ PLUS membrane for fastest flow rates and low protein binding
- Larger scale, pressure-driven filtration device for faster flow rates speeds time-consuming, large-volume filtrations
- 10 L and 20 L capacity sizes for maximum laboratory-scale filtration options
- Manually operated top air vent to prevent air lock stop of flow
- Filling bell for cleaner filling environment around the opening of the receptacle bottle
Applications:
Sterilization of Aqueous Solutions, Media Preparation, Buffer Preparation
Specifications
Single-use | |
PES | |
Sterile | |
3Pack | |
6mm stepped hose barb, filling bell | |
45°C | |
2.99 in. | |
9.9 cm |
SAN | |
0.22 μm | |
75 psi forward, 5 psi reverse | |
6 mm stepped hose barb, filling bell | |
113°F | |
Bottle Top Filter | |
7.6 cm |