Learn More
Brady™ Engraved Replacement Labels for M610 and M611
Engraved Plate Replacement Labels for use with Brady™ Printers M611 and M610.
Brand:  Brady™ 312031
 View more versions of this product
Product Code. 17848054
  • £242.00 / Pack of 300
Estimated Shipment: 13-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Engraved Plate Replacement labels for the M610 Portable Printer
  • Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags
  • Service temperature of -40°C to +100°C
Specifications
Specifications
Permanent Acrylic, Foam-backed
Yellow
Label
Single-side Printable
Engraved Plate Labels
Data and Telecommunications, Electrical Labeling, General Identification
1 Label
300 Labels
-40°F to +212°F
0.0256 in.
45 x 15 mm
0.59 in.
15 mm
None
Gloss
Polyester
Thermal Transfer
Non-sterile
M610, M611, BMP61
-40°C
Rectangle
-40°C to +100°C
0.65 mm
1.77 in.
15 mm