Learn More
Sartorius Entris™ II Advanced Analytical Balances with Country-Specific Verification
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
£3340.00 - £4140.00
Specifications
Certifications/Compliance CE Certificate
Connection Type D-sub connector, 9-pin; USB UTL; USB Device
Display Type Intuitive Graphic Touch Technology
Readability 0.1 mg
Material Glass, PBT, Stainless Steel
View More Specs
Products 4
Product Code Brand Capacity Description Price Quantity & Availability  
Product Code Brand Capacity Description Price Quantity & Availability  
16685683
View accessory products View Documents
Sartorius
BCA64I-1CEU
60 g Verified
£3340.00
Each
16626243
View accessory products View Documents
Sartorius
BCA124I-1CEU
120 g Verified
£3430.00
Each
16635653
View accessory products View Documents
Sartorius
BCA224I-1CEU
220 g Verified
£3700.00
Each
16626253
View accessory products View Documents
Sartorius
BCA324I-1CEU
320 g Verified
£4140.00
Each
Description
Description
  • Entris™ II Advanced balances - Quality, Value, and Consistency
  • For accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in real-time level support
  • Increases reliability of weighing results using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail
Specifications
Specifications
CE Certificate
Intuitive Graphic Touch Technology
Glass, PBT, Stainless Steel
≤1.5 sec.
0.2 A
D-sub connector, 9-pin; USB UTL; USB Device
0.1 mg
Electronic Analytical Balance
100/240 V (±10%)
50/60 Hz
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Sartorius Entris™ II Advanced Analytical Balances with Country-Specific Verification