Learn More
Memmert™ Entry Port 23 mm, Flap Closure, Custom Position Tbc In The Back For Un75Pa
Use these entry ports with convection ovens. Memmert™ Convection and Universal Oven Entry Points can be closed by flap in special positions at the back.
Brand:  Memmert™ UN75paF6
Product Code. 16344497
  • £244.00 / Each
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Entry port with clear diameter
  • For Memmert Ovens
Specifications
Specifications
Introducing Moisture Tight Connections, Can be Closed by Flap in Special Rear Positions
Entry Port
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Entry Port 23 mm, Flap Closure, Custom Position Tbc In The Back For Un75Pa >