missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Vision™ Salmonella Slide Agglutination Antisera

A comprehensive range of Salmonella slide agglutination antisera

Brand:  Pro-Lab Diagnostics™ PL.6015

 View more versions of this product

Product Code. 11909064

  • £220.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • All sera produced in conjunction with the Salmonella reference section of the Veterinary Laboratory Agency, Weybridge, UK
  • Range includes a full panel of polyvalent somatic antisera, monovalent somatic antisera, polyvalent flagella antisera and monovalent flagella antisera for traditional slide agglutination testing
  • Shelf life up to five years from date of manufacture

Order Info

Supplied in color-coded 3.0mL dropper bottles

Specifications

Specifications

2°C to 8°C
Genus Salmonella
Antisera Slide
3 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pro-Lab Diagnostics™ Vision™ Salmonella Slide Agglutination Antisera > Monovalent Somatic; Group D; Factor 9

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.