missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Sterile Plastic Inoculation Loops (Volume: 10μL)
Retrieves an inoculum from culture of microorganisms.
Brand: Microspec™ A4G
Description
- High Impact Polystyrene construction
- Full length to avoid contamination
- Disposable
- Sterile, aseptically manufactured
- Medium rigidity
- Culture loops (Length: 200mm)
- Smooth edges to avoid tearing and digging in agar
- Volume: 10μL
- Color: Blue
Specifications
Single-use | |
0.18 in. | |
7.87 in. | |
High-impact Polystyrene | |
Blue | |
7.87 in. | |
50 Pouches of 20 | |
Yes |
10 μL | |
4.5 mm | |
200 mm | |
Gamma-irradiated | |
Inoculating Product | |
200 mm | |
Microseal Pouch |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Microspec™ Sterile Plastic Inoculation Loops (Volume: 10μL) > 1000/Pk.; Microseal pack of 20, Gamma irradiated
Spot an opportunity for improvement?Share a Content Correction