Learn More
Waters Corporation Envirogel GPC Cleanup Column
The Envirogel High-Resolution GPC cleanup columns are specifically designed to remove low volatility, high molecular weight interferences.
Brand:  Waters Corporation 186001916
Encompass
 View more versions of this product
Product Code. 13127669
  • £4998.00 / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • In the past, the cleanup procedure for environmental samples was performed on low-efficiency GPC columns based on packing particle diameters of 37-75 micron (200-400 mesh) Biobeads S-X resins.
  • The high-efficiency Envirogel Cleanup columns increase the speed of this process while simultaneously reducing solvent consumption.
  • They come packed in your choice of Methylene Chloride or a mixture of Cyclohexane/Ethyl acetate to minimize the need for risky solvent switchover.
Specifications
Specifications
19 mm
Spherical
1/Ea.
30 mm
15 μm
Column