missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation Envirogel GPC Cleanup Column
The Envirogel High-Resolution GPC cleanup columns are specifically designed to remove low volatility, high molecular weight interferences.
£3597.00 - £4998.00
Specifications
Diameter (Metric) | 19 mm |
---|---|
Particle Shape | Spherical |
Particle Size | 15 μm |
Quantity | 1/Ea. |
Type | Column |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
13117669
|
Waters Corporation
186001915 |
150 mm |
£3597.00
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||||
13127669
|
Waters Corporation
186001916 |
30 mm |
£4998.00
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||||
Description
- In the past, the cleanup procedure for environmental samples was performed on low-efficiency GPC columns based on packing particle diameters of 37-75 micron (200-400 mesh) Biobeads S-X resins.
- The high-efficiency Envirogel Cleanup columns increase the speed of this process while simultaneously reducing solvent consumption.
- They come packed in your choice of Methylene Chloride or a mixture of Cyclohexane/Ethyl acetate to minimize the need for risky solvent switchover.
Specifications
19 mm | |
15 μm | |
Column |
Spherical | |
1/Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title