Learn More
Thermo Scientific Chemicals Eosin B, pure, high purity, biological stain
Eosin B, C20H6Br2N2Na2O9, CAS Number-548-24-3, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, 4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, Eosin Bluish, 100g, 0.96 to 1.22, 19, 233, Brown, 208-943-1, 624.06, Pure, Danger, GHS H Statement: Causes serious eye damage.
£30.70 - £60.40
Chemical Identifiers
CAS | 548-24-3 |
---|---|
Molecular Formula | C20H6Br2N2Na2O9 |
MDL Number | MFCD00005041 |
Synonym | 4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, Eosin Bluish |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10174660
|
Thermo Scientific Chemicals
229650250 |
25 g | Glass bottle |
£30.70
25g |
In Stock
Log in to see stock. |
||||
10086183
|
Thermo Scientific Chemicals
229651000 |
100 g | Glass bottle |
£60.40
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
548-24-3 | |
C20H6Br2N2Na2O9 | |
MFCD00005041 | |
4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, Eosin Bluish |
Specifications
548-24-3 | |
C20H6Br2N2Na2O9 | |
19, 233 | |
4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, Eosin Bluish | |
511 to 520nm (in water + 1% Na2CO3) | |
Pure | |
Crystalline Powder |
0.96 to 1.22 | |
MFCD00005041 | |
15, 3657 | |
Authentic | |
624.06 | |
Brown | |
Eosin B |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 208-943-1
missing translation for 'tsca' : TSCA
RUO – Research Use Only