missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD

The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For In Vitro Diagnostic Use.

Brand:  Agilent BioTek EPOCH2NSC-SI


Additional Details : Weight : 11.00000kg

 View more versions of this product

Product Code. 17237809

  • POA / Each
Estimated Shipment: 14-08-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »


Description

Description

The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. Measurements can be performed in 6- to 384-well microplates, cuvets, and in microvolume samples with the available Take3 microvolume plate. The broad wavelength range enables applications from nucleic acid and protein quantification in the low UV to microbial growth assays higher wavelengths

Epoch 2 is easy to use and the optional touch screen interface makes it simple to choose predefined protocols or to define custom programs. Endpoint, kinetic, spectral scanning and well area scanning modes, plus incubation and shaking enable wide-ranging applications.

Specifications

Specifications

Photodiode
96 wells: 8 sec., 384 wells: 14 sec.
CE and TUV marked. RoHS compliant.
22 in.
Cuvet port only
200 to 999 nm, 1 nm increments
Xenon Flash Lamp
±2 nm
Nucleic acid and protein quantification to ELISA and microbial growth assays
50/60 Hz
EPOCH2NSC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers