missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Epoxy Diagnostic Slides
These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use
Brand: Epredia™ X1XER308B
141.75 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Binds and keeps liquids applied to the well area
- Provides wettable wells for staining and growing cell cultures
- Prevents cross-contamination
- Hydrophobic coatings are very durable and withstand the strongest stains
- Autoclavable
Specifications
25 mm | |
Epoxy | |
Diagnostic Slide | |
Black |
75 mm | |
1 mm | |
10 | |
6.7 mm Well |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction