Learn More
Eppendorf epT.I.P.S.™ Motion as SafeRack tips
GREENER_CHOICE
The new epMotion systems can reuse tips as e.g. economical choice for multiple wash steps of the same well.
Brand:  Eppendorf 0030014618
207.99 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
 View more versions of this product
Product Code. 30019050
  • £226.00 / Pack of 960
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Specially designed for use on the epMotion™ 5070 and 5075 Liquid Handling Workstation. epT.I.P.S. Motion SafeRack pipette tips, with and without filters
  • Developed in torsion-resistant, contamination-free racks that are recommended for the re-use of tips feature of epMotion 5073 systems.
Reuse Tips Feature:
  • The new epMotion systems can reuse tips as e.g. economical choice for multiple wash steps of the same well
  • Pipette tips will be placed back into the position of the box they were removed from
  • Returned tips will be reused within an application for the same source vessel
  • Individual compartments in rack to avoid cross-contamination between used tips
  • Available as standard and PCR clean quality for 50, 300, and 1000 μL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 μL
PCR Clean
Not Autoclavable
Box
Robotic Tips
Eppendorf epMotion
Polypropylene
Filtered
Clear
For Eppendorf epMotion series
Standard, Straight
Single-use
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf epT.I.P.S.™ Motion as SafeRack tips > Volume (Metric): 50 μL; Filtered: Filtered; Sterility: PCR clean