missing translation for 'onlineSavingsMsg'
Learn More
Erbium foil, 0.62mm (0.024in) thick, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11326587

Erbium foil, 0.62mm (0.024in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11326587
25 x 25 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy

Product Code. 11326587

Brand: Thermo Scientific Alfa Aesar 000593.FF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-52-0
Molecular Formula Er
Molecular Weight (g/mol) 167.26
MDL Number MFCD00010987
InChI Key UYAHIZSMUZPPFV-UHFFFAOYSA-N
Synonym unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
PubChem CID 23980
ChEBI CHEBI:33379
IUPAC Name erbium
SMILES [Er]

Specifications

Quantity 25 x 25 mm
Percent Purity 99.9%
Odor Odorless
Weight ≈3.5g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Erbium foil, 0.62mm (0.024 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberK02W041 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000593.FI,000593.FF
Lot NumberE06T033 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000593.FI,000593.FF
Lot NumberN17B049 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000593.FI,000593.FF
Lot NumberE20Q25 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000593.FI,000593.FF
Lot NumberL18R026 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000593.FI,000593.FF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Erbium foil, 0.62mm (0.024in) thick, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.