missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

£84.72 - £264.32

Chemical Identifiers

CAS 19423-85-9
Molecular Formula Cl3Er
Molecular Weight (g/mol) 273.61
MDL Number MFCD00010989
InChI Key HDGGAKOVUDZYES-UHFFFAOYSA-K
IUPAC Name erbium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Er+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11351049
View Documents
Thermo Scientific Alfa Aesar
011304.14
25 g
£84.72
25g
Please to purchase this item. Need a web account? Register with us today!
11341049
View Documents
Thermo Scientific Alfa Aesar
011304.22
100 g
£264.32
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Erbium(III) chloride hydrate is used as a starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18, for which electron and stability studies were performed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19423-85-9
273.61
HDGGAKOVUDZYES-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[Er+3]
Cl3Er
MFCD00010989
erbium(3+) trichloride

Specifications

19423-85-9
Cl3Er
Slightly soluble in ethanol
[Cl-].[Cl-].[Cl-].[Er+3]
273.61
99.9%
Odorless
(REO)
Crystalline Aggregates
MFCD00010989
HDGGAKOVUDZYES-UHFFFAOYSA-K
erbium(3+) trichloride
273.62 (Anhydrous)
Reacton™
Hygroscopic
Erbium(III) chloride hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 233-385-0

missing translation for 'rtecsNumber' : KD8575000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only