Learn More
Erbium(III) perchlorate, 40 wt.% solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 428290050
43.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Erbium(III) perchlorate | |
5 g | |
MFCD00016073 | |
FGBKFKCLWFRKEW-UHFFFAOYSA-K | |
erbium(3+);triperchlorate | |
15335705 | |
Glass bottle | |
1.4400g/mL |
14017-55-1, 7732-18-5 | |
Cl3ErO12 | |
erbium 3+ perchlorate, erbium perchlorate, perchloric acid,erbium 3+ salt 8ci,9ci, erbium iii perchlorate solution, perchloric acid erbium iii salt, erbium perchlorate aqueous solution | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3] | |
465.61 | |
465.61 | |
1.44 |
Safety and Handling
- Erbium(III) perchlorate, 40 wt.% solution in water
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0397126 | Certificate of Analysis | 14/12/2022 | 428290250,428290050 |
A0406797 | Certificate of Analysis | 13/12/2022 | 428290250,428290050 |
A0405557 | Certificate of Analysis | 13/12/2022 | 428290250,428290050 |
A0416914 | Certificate of Analysis | 26/08/2022 | 428290250,428290050 |
A0440838 | Certificate of Analysis | 20/04/2022 | 428290250,428290050 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only