missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion™ Hot Start II DNA Polymerases (2 U/μL)

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR.

Brand:  Thermo Scientific™ F537L

 View more versions of this product

Product Code. 13595160

  • £580.00 / 500 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes:

Phusion Hot Start II High–Fidelity DNA Polymerase

  • Phusion Hot Start II High–Fidelity DNA Polymerase (2 U/μL)
  • 5X Phusion HF Buffer
  • 5X Phusion GC Buffer
  • DMSO
  • 50 mM MgCl2 solution

Phusion Green Hot Start II High–Fidelity DNA Polymerase

  • Phusion Hot Start II High–Fidelity DNA Polymerase (2 U/Glc μL)
  • 5X Phusion Green HF Buffer
  • 5X Phusion Green GC Buffer
  • DMSO
  • 50 mM MgCl2 solution

Both Phusion and Phusion Green Buffer provide 1.5 mM MgCl2 in the final 1X concentration

This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• Phusion Hot Start II High-Fidelity DNA Polymerase (2 U/μL)
• 5X Phusion Green HF Buffer
• 5X Phusion Green GC Buffer
• DMSO
• 50 mM MgCl2 solution

Phusion Green HF and GC Buffers both provide 1.5 mM MgCl2 in the final 1X concentration.
Store at –20°C.
Phusion Hot Start II High-Fidelity DNA Polymerase
Hot Start High-Fidelity DNA Polymerase
Dry Ice
2 U/μL
Built-In Hot Start
Blunt
20 kb or less
High
Fast
500 units
Hot-start PCR, High-fidelity PCR
52X
500 Reactions
Standalone
Green
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2830617 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberF537L
Lot Number2870108 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberF537L
Lot Number2888197 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberF537L
Lot Number2922231 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberF537L
Lot Number3069416 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberF537L
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Phusion™ Hot Start II DNA Polymerases (2 U/μL) > Phusion™ Green Hot Start II High-Fidelity DNA Polymerase (2 U/μL), 500 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.