Learn More
Velp Scientifica™ ESP Ultraflat Magnetic Stirrer
ESP Ultraflat Magnetic Stirrer with gentle start-up ensures optimum progression of the stirring speed whilst its modern, ergonomic structure is made of materials that ensure a high resistance to chemical reagents.
Brand: Velp Scientifica™ F206A0179
166.59 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Description
Durable Design:- Maintenance-free: contains no moving parts
- Control panel is protected from liquid spills with run-off groove
- Technopolymer casing resists chemicals
- Long life operation
- Accepts up to 5 liter flasks
- Surface ideal for visualization of particles and color changes
- Ideal for microtitration, microbiology and biochemistry
- Soft start but rapidly reaching set speed
- Ergonomic - very low height
- Speed regulation up to 1100 rpm
- Universal mains voltage / frequency range
- Light weight
Specifications
1100 rpm | |
Technopolymer | |
Analog | |
5 w | |
33 mm | |
160 mm | |
50/60 Hz | |
Magnetic Stirrer |
1 | |
5 L | |
100/240 V | |
230 x 160 x 33 mm | |
230 mm | |
100/240 V, 50/60 Hz, 5 W | |
ESP | |
0.9 kg |
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.