missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Estradiol 3-methyl ether solution, Honeywell Fluka™
100 μg/mL in acetonitrile, analytical standard, reference material
Brand: Honeywell Fluka R1357-2ML
This item is not returnable.
View return policy
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support

Specifications
75-05-8, 1035-77-4 | |
2 mL | |
Steroids and Hormones | |
82°C |
Glass Bottle | |
6°C | |
Liquid | |
Analytical Standard |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
β-Estradiol 3-methyl ether solution, Honeywell Fluka™ > 2mL
Spot an opportunity for improvement?Share a Content Correction