Learn More
Estrone, MedChemExpress
MedChemExpress Estrone (E1) is a natural estrogenic hormone. Estrone is the main representative of the endogenous estrogens and is produced by several tissues, especially adipose tissue. Estrone is the result of the process of aromatization of androstenedione that occurs in fat cells.
Brand: Medchem Express HY-B0234
Description
Estrone (E1) is a natural estrogenic hormone. Estrone is the main representative of the endogenous estrogens and is produced by several tissues, especially adipose tissue. Estrone is the result of the process of aromatization of androstenedione that occurs in fat cells.Specifications
Estrone | |
White | |
C18H22O2 | |
E1 Oestrone | |
C[C@]1([C@](CC2)([H])[C@]3([H])CCC4=C(C=CC(O)=C4)[C@@]3([H])CC1)C2=O | |
270.37 | |
Research | |
Cancer-programmed cell death |
53-16-7 | |
Research | |
5 g | |
DMSO : 25 mg/mL (92.47 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 0.1 mg/mL (0.37 mM; Need ultrasonic) | |
270.37 | |
97.0% | |
Powder |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only