Learn More
Estrone, MedChemExpress
MedChemExpress Estrone (E1) is a natural estrogenic hormone. Estrone is the main representative of the endogenous estrogens and is produced by several tissues, especially adipose tissue. Estrone is the result of the process of aromatization of androstenedione that occurs in fat cells.
£75.02 - £211.75
Specifications
Chemical Name or Material | Estrone |
---|---|
CAS | 53-16-7 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C18H22O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16478428
|
Medchem Express
HY-B0234 |
10 g |
£211.75
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16460838
|
Medchem Express
HY-B0234 |
5 g |
£128.26
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16412109
|
Medchem Express
HY-B0234 |
1 g |
£75.02
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Estrone (E1) is a natural estrogenic hormone. Estrone is the main representative of the endogenous estrogens and is produced by several tissues, especially adipose tissue. Estrone is the result of the process of aromatization of androstenedione that occurs in fat cells.Specifications
Estrone | |
White | |
C18H22O2 | |
DMSO : 25 mg/mL (92.47 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 0.1 mg/mL (0.37 mM; Need ultrasonic) | |
270.37 | |
97.0% | |
Powder |
53-16-7 | |
Research | |
E1 Oestrone | |
C[C@]1([C@](CC2)([H])[C@]3([H])CCC4=C(C=CC(O)=C4)[C@@]3([H])CC1)C2=O | |
270.37 | |
Research | |
Cancer-programmed cell death |
Safety and Handling
H225∣H301+H311+H331∣H370
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.