Learn More
Estropipate, MedChemExpress
MedChemExpress Estropipate is a form of estrogen, used to treat symptoms of menopause, also used to prevent osteoporosis.
Brand: Medchem Express HY-B1361
This item is not returnable.
View return policy
Description
Estropipate is a form of estrogen, used to treat symptoms of menopause, also used to prevent osteoporosis.Specifications
Estropipate | |
White | |
C22H32N2O5S | |
Piperazine estrone sulfate Estrone sulfate piperazine salt | |
C[C@]1([C@](CC2)([H])[C@]3([H])CCC4=C(C=CC(OS(=O)(O)=O)=C4)[C@@]3([H])CC1)C2=O.N5CCNCC5 | |
436.56 | |
Research |
7280-37-7 | |
Research | |
10 mg | |
DMSO : 110 mg/mL (251.97 mM; Need ultrasonic) | |
436.56 | |
99.0% | |
Solid |