missing translation for 'onlineSavingsMsg'
Learn More

Estropipate, MedChemExpress

MedChemExpress Estropipate is a form of estrogen, used to treat symptoms of menopause, also used to prevent osteoporosis.

£56.87 - £114.95

Specifications

Chemical Name or Material Estropipate
CAS 7280-37-7
Color White
Purity Grade Notes Research
Molecular Formula C22H32N2O5S
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16444587
Encompass_Preferred
View Documents
Medchem Express
HY-B1361
50 mg
£114.95
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16428698
Encompass_Preferred
View Documents
Medchem Express
HY-B1361
10 mg
£56.87
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Estropipate is a form of estrogen, used to treat symptoms of menopause, also used to prevent osteoporosis.
Specifications

Specifications

Estropipate
White
C22H32N2O5S
DMSO : 110 mg/mL (251.97 mM; Need ultrasonic)
436.56
99.0%
Solid
7280-37-7
Research
Piperazine estrone sulfate Estrone sulfate piperazine salt
C[C@]1([C@](CC2)([H])[C@]3([H])CCC4=C(C=CC(OS(=O)(O)=O)=C4)[C@@]3([H])CC1)C2=O.N5CCNCC5
436.56
Research
Videos
Safety and Handling

Safety and Handling

H302∣H350

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.