missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1-Boc-3-benzylpiperidine-3-carboxylate, 95%, Thermo Scientific™
£30.14 - £90.10
Chemical Identifiers
CAS | 170842-80-5 |
---|---|
Molecular Formula | C20H29NO4 |
Molecular Weight (g/mol) | 347.46 |
MDL Number | MFCD11500859 |
InChI Key | ZTGGNYIHGLOCJH-UHFFFAOYNA-N |
Synonym | 1-tert-butyl 3-ethyl 3-benzylpiperidine-1,3-dicarboxylate, ethyl 1-boc-3-benzylpiperidine-3-carboxylate, 3-benzylpiperidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-ethyl ester, +/--n-t-butoxycarbonyl-3-carboethoxy-3-phenylmethyl-piperidine, 3-benzylpiperidine-1,3-dicarboxylic acid1-tert-butyl ester 3-ethyl ester, 1,3-piperidinedicarboxylic acid, 3-phenylmethyl-, 1-1,1-dimethylethyl 3-ethyl ester, 1,3-piperidinedicarboxylicacid,3-phenylmethyl-,1-1,1-dimethylethyl 3-ethylester |
PubChem CID | 19696502 |
IUPAC Name | 1-O-tert-butyl 3-O-ethyl 3-benzylpiperidine-1,3-dicarboxylate |
SMILES | CCOC(=O)C1(CC2=CC=CC=C2)CCCN(C1)C(=O)OC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458715
|
Thermo Scientific Alfa Aesar
H33958.MD |
250 mg | N/A | ||||||
15486847
|
Thermo Scientific Alfa Aesar
H33958.03 |
1 g |
£90.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
170842-80-5 | |
347.46 | |
ZTGGNYIHGLOCJH-UHFFFAOYNA-N | |
19696502 | |
CCOC(=O)C1(CC2=CC=CC=C2)CCCN(C1)C(=O)OC(C)(C)C |
C20H29NO4 | |
MFCD11500859 | |
1-tert-butyl 3-ethyl 3-benzylpiperidine-1,3-dicarboxylate, ethyl 1-boc-3-benzylpiperidine-3-carboxylate, 3-benzylpiperidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-ethyl ester, +/--n-t-butoxycarbonyl-3-carboethoxy-3-phenylmethyl-piperidine, 3-benzylpiperidine-1,3-dicarboxylic acid1-tert-butyl ester 3-ethyl ester, 1,3-piperidinedicarboxylic acid, 3-phenylmethyl-, 1-1,1-dimethylethyl 3-ethyl ester, 1,3-piperidinedicarboxylicacid,3-phenylmethyl-,1-1,1-dimethylethyl 3-ethylester | |
1-O-tert-butyl 3-O-ethyl 3-benzylpiperidine-1,3-dicarboxylate |
Specifications
170842-80-5 | |
MFCD11500859 | |
ZTGGNYIHGLOCJH-UHFFFAOYNA-N | |
1-O-tert-butyl 3-O-ethyl 3-benzylpiperidine-1,3-dicarboxylate | |
19696502 | |
95% |
C20H29NO4 | |
1-tert-butyl 3-ethyl 3-benzylpiperidine-1,3-dicarboxylate, ethyl 1-boc-3-benzylpiperidine-3-carboxylate, 3-benzylpiperidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-ethyl ester, +/--n-t-butoxycarbonyl-3-carboethoxy-3-phenylmethyl-piperidine, 3-benzylpiperidine-1,3-dicarboxylic acid1-tert-butyl ester 3-ethyl ester, 1,3-piperidinedicarboxylic acid, 3-phenylmethyl-, 1-1,1-dimethylethyl 3-ethyl ester, 1,3-piperidinedicarboxylicacid,3-phenylmethyl-,1-1,1-dimethylethyl 3-ethylester | |
CCOC(=O)C1(CC2=CC=CC=C2)CCCN(C1)C(=O)OC(C)(C)C | |
347.46 | |
347.45 | |
Ethyl 1-Boc-3-benzylpiperidine-3-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures