missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-(4-pyridyl)thiazole-4-acetate, 97%, Thermo Scientific™
£12.00 - £135.00
Chemical Identifiers
CAS | 80653-68-5 |
---|---|
Molecular Formula | C12H12N2O2S |
Molecular Weight (g/mol) | 248.3 |
MDL Number | MFCD06739821 |
InChI Key | JZBJUBBYJUUKQK-UHFFFAOYSA-N |
Synonym | ethyl 2-2-pyridin-4-yl thiazol-4-yl acetate, ethyl 2-4-pyridyl thiazole-4-acetate, 2-pyridin-4-yl-thiazol-4-yl-acetic acid ethyl ester, 2-4-pyridyl thiazole-4-acetic acid ethyl ester, ethyl 2-2-pyridin-4-yl-1,3-thiazol-4-yl acetate, 2-pyridin-4-yl-thiazol-4-yl-acetic acid ethylester |
PubChem CID | 7176350 |
IUPAC Name | ethyl 2-(2-pyridin-4-yl-1,3-thiazol-4-yl)acetate |
SMILES | CCOC(=O)CC1=CSC(=N1)C2=CC=NC=C2 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
80653-68-5 | |
248.3 | |
JZBJUBBYJUUKQK-UHFFFAOYSA-N | |
7176350 | |
CCOC(=O)CC1=CSC(=N1)C2=CC=NC=C2 |
C12H12N2O2S | |
MFCD06739821 | |
ethyl 2-2-pyridin-4-yl thiazol-4-yl acetate, ethyl 2-4-pyridyl thiazole-4-acetate, 2-pyridin-4-yl-thiazol-4-yl-acetic acid ethyl ester, 2-4-pyridyl thiazole-4-acetic acid ethyl ester, ethyl 2-2-pyridin-4-yl-1,3-thiazol-4-yl acetate, 2-pyridin-4-yl-thiazol-4-yl-acetic acid ethylester | |
ethyl 2-(2-pyridin-4-yl-1,3-thiazol-4-yl)acetate |
Specifications
80653-68-5 | |
MFCD06739821 | |
JZBJUBBYJUUKQK-UHFFFAOYSA-N | |
ethyl 2-(2-pyridin-4-yl-1,3-thiazol-4-yl)acetate | |
7176350 | |
97% |
C12H12N2O2S | |
ethyl 2-2-pyridin-4-yl thiazol-4-yl acetate, ethyl 2-4-pyridyl thiazole-4-acetate, 2-pyridin-4-yl-thiazol-4-yl-acetic acid ethyl ester, 2-4-pyridyl thiazole-4-acetic acid ethyl ester, ethyl 2-2-pyridin-4-yl-1,3-thiazol-4-yl acetate, 2-pyridin-4-yl-thiazol-4-yl-acetic acid ethylester | |
CCOC(=O)CC1=CSC(=N1)C2=CC=NC=C2 | |
248.3 | |
248.3 | |
Ethyl 2-(4-pyridyl)thiazole-4-acetate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures